Learn More
Chemglass Life Sciences Adapter, Connecting, 24/40 Joint Size, Valve, Airfree, Schlenk
SDP
Catalog No. AF050016
Encompass_Preferred
Adapter, Connecting, 24/40 Joint Size, Valve, Airfree, Schlenk. Converts standard flasks and other apparatus to Airfree applications. With top outer and lower inner standard taper joints, both being the same size. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retention device or a 0-4mm Chem-Cap Airfree inlet valve. This item comes with a Chem-Cap Airfree inlet valve.
Catalog No. AF050016 Supplier Chemglass Life Sciences Supplier No. AF050016
Adapter, Connecting, 24/40 Joint Size, Valve, Airfree, Schlenk. Converts standard flasks and other apparatus to Airfree applications. With top outer and lower inner standard taper joints, both being the same size. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retention device or a 0-4mm Chem-Cap Airfree inlet valve. This item comes with a Chem-Cap Airfree inlet valve.