Learn More
Chemrus Inc ADAPTER 24-400 THREAD JOINT 1
Catalog No. 5082320
Encompass
Chemrus glass vacuum adapter, glass taper outer joint on the top to connect disposable filter funnels, PTFE stopcock to control the vacuum source, fit 24-400 thread joint
Catalog No. 50-823-20 Supplier Chemrus Inc Supplier No. CR202804
Chemrus glass vacuum adapter, glass taper outer joint on the top to connect disposable filter funnels, PTFE stopcock to control the vacuum source, fit 24-400 thread joint