Learn More
Beckman Coulter Beckman Coulter adapter for RIA tubes, 25 x 12 x 75 mm, for SX4250 swing-bucket rotor (set of 2)
Catalog No. NC2462641
Encompass_Preferred
Adapter set designed to hold 25 RIA tubes (12 x 75 mm) and adapt them for use in swinging-bucket rotors. Sold as a set of two adapters, this accessory supports routine centrifugation of 12 mm RIA-style tubes.
  • Holds 25 RIA tubes.
  • Accommodates tubes 12 x 75 mm (diameter x length).
  • Sold as a set of two adapters.
  • Compatible with swinging-bucket rotors such as the SX4250.
Catalog No. NC2462641 Supplier Beckman Coulter Supplier No. 392263
Adapter set designed to hold 25 RIA tubes (12 x 75 mm) and adapt them for use in swinging-bucket rotors. Sold as a set of two adapters, this accessory supports routine centrifugation of 12 mm RIA-style tubes.
  • Holds 25 RIA tubes.
  • Accommodates tubes 12 x 75 mm (diameter x length).
  • Sold as a set of two adapters.
  • Compatible with swinging-bucket rotors such as the SX4250.