Learn More
Ace Glass, Inc. Adapter, 28/15 socket joint, #15 Ace-Thred end
SDP
Catalog No. NC9758545
Encompass_Preferred
28/15 socket joint adapter with #15 Ace-Thred end, for joining jacketed reactor ball joint inlet/outlets to recirculator hoses.
Catalog No. NC9758545 Supplier Ace Glass, Inc. Supplier No. 522111
28/15 socket joint adapter with #15 Ace-Thred end, for joining jacketed reactor ball joint inlet/outlets to recirculator hoses.