Learn More
Chemglass Life Sciences 1/4" Thermocouple Adapter, 29/42 Inner
SDP
Supplier:  Chemglass Life Sciences CG1042E07
Encompass_Preferred
1/4" Thermocouple Adapter, 29/42 Inner. Adapter has a Chem-Thread at the top to make a vacuum tight seal. It will accept our 1/4in OD thermocouple probes. Lower portion of the adapter is reduced down to hold the thermocouple securely in place and away from the agitators. Supplied complete with a Celcon cap and Viton o-ring. Perfluoro o-rings are available.
Catalog No. CG1042E07