Learn More
Ace Glass, Inc. Adapter, tee tube compression, 29/42, 3/4in OD ground glass sidearm
SDP
Catalog No. 50977833
Encompass_Preferred
Catalog No. 50-977-833 Supplier Ace Glass, Inc. Supplier No. 1273720