Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Adapter with 3/4 in. NPT Thread x 5/8 in. Barb - Natural Kynar
SDP

Catalog No. 01000075 Shop All Fisherbrand Products
Change view
Click to view available options
Packaging Quantity:
10 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
01-000-075 10 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-000-075
Add to Cart
Add to Cart
This item is not returnable. View return policy

Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing.

  • Single & Multi Use Applications
  • PVC-Free
  • Superior Single Barb Design
  • USP Class VI Compliant
  • USP 661
  • USP81
  • USP 8
  • RoHS Compliant
  • Gamma Stable
  • Animal Derivative Free
  • Lot Traceable

Warranty and Services

Replacement if not satisfied. 12 Month return Policy.

Specifications

Certifications/Compliance Compliance Certificates Available
Material Kynar PVDF
For Use With (Application) Threaded Tube Connector for Medical, Pharmaceutical, Industrial applications.
Packaging Quantity 10 Pack
Storage Requirements Out of direct sunlight, >20°F <100°F
Color Natural
Product Type Threaded Adapter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.