Learn More
Chemglass Life Sciences Adapter, 3/4" Beaded Pipe to 3/4" Compression Fitting, Glass Filled TFE
SDP
Catalog No. 501531309
Encompass_Preferred
Adapter, 3/4" Beaded Pipe to 3/4" Compression Fitting. Made from Glass Filled TFE. Glass filled PTFE adapter for use with the CG-1968-P zero dead space detachable valve. Adapts 1" beaded pipe to a 3/4" compression fitting for use with CG-1975 adapters and tubing. Requires a CG-1968-66 1" beaded pipe coupling.
Catalog No. 50-153-1309 Supplier Chemglass Life Sciences Supplier No. CG196450A
Adapter, 3/4" Beaded Pipe to 3/4" Compression Fitting. Made from Glass Filled TFE. Glass filled PTFE adapter for use with the CG-1968-P zero dead space detachable valve. Adapts 1" beaded pipe to a 3/4" compression fitting for use with CG-1975 adapters and tubing. Requires a CG-1968-66 1" beaded pipe coupling.