Learn More
Julabo USA Inc Pair, M16x1f to 3/8" tube stub adapters
SDP
Catalog No. 507534191
Encompass_Preferred
Bath Accessories, Julabo, Adapter 3/8in. tube, Pair of M16x1 female to 3/8" tube stub adapters. For use with Swagelok union fittings.
Catalog No. 50-753-4191 Supplier Julabo USA Inc Supplier No. 8890012
Bath Accessories, Julabo, Adapter 3/8in. tube, Pair of M16x1 female to 3/8" tube stub adapters. For use with Swagelok union fittings.