Learn More
Grainger Adapter 316 SS 2 x 2 in Pipe Size Socket
Catalog No. 502748690
Encompass_Preferred
The Blucher Stainless Steel Adapter is a sanitary fitting used to connect tubes, pipes, and hoses in food, beverage, pharmaceutical, and other hygienic processing systems. It is made from 316L stainless steel for corrosion resistance. This tri-clamp adapter has a 1-1/2" sanitary clamp on one end and a 1-1/2" sanitary clamp on the other end. The maximum pressure rating is 232 psi at 70°F. The maximum temperature rating is 250°F. The adapter body height is 2-3/4". It is manufactured in compliance with 3-A sanitary standards.
Catalog No. 50-274-8690 Supplier Grainger Supplier No. 14P011
The Blucher Stainless Steel Adapter is a sanitary fitting used to connect tubes, pipes, and hoses in food, beverage, pharmaceutical, and other hygienic processing systems. It is made from 316L stainless steel for corrosion resistance. This tri-clamp adapter has a 1-1/2" sanitary clamp on one end and a 1-1/2" sanitary clamp on the other end. The maximum pressure rating is 232 psi at 70°F. The maximum temperature rating is 250°F. The adapter body height is 2-3/4". It is manufactured in compliance with 3-A sanitary standards.