Learn More
Ace Glass, Inc. Adapter, vertical tube compression, 35/25 socket, 3/4in glass sidearm
SDP
Catalog No. 50977805
Encompass_Preferred
Catalog No. 50-977-805 Supplier Ace Glass, Inc. Supplier No. 1273148