Learn More
Chemglass Life Sciences Adapter, 45/50 Inner to 1" Sanitary, 316L Stainless Steel
SDP
Catalog No. 501310986
Encompass_Preferred
Adapter, 45/50 Inner to 1" Sanitary, 316L Stainless Steel. Standard taper inner joint to sanitary fitting adapter. Provides a convenient way to connect sanitary fittings and adapters to Chemglass reaction systems. Approximate overall length is 4¿ (102mm). NOTE: Clamp and Seals Must Be Purchased Separately.
Catalog No. 50-131-0986 Supplier Chemglass Life Sciences Supplier No. CG1002B20
Adapter, 45/50 Inner to 1" Sanitary, 316L Stainless Steel. Standard taper inner joint to sanitary fitting adapter. Provides a convenient way to connect sanitary fittings and adapters to Chemglass reaction systems. Approximate overall length is 4¿ (102mm). NOTE: Clamp and Seals Must Be Purchased Separately.