Learn More
Chemglass Life Sciences Adapter, 45/50 Inner to 1" Sanitary, Glass Filled PTFE
SDP
Catalog No. 5000980718
Encompass_Preferred
Used to connect dissimilar standard taper joint sizes. Lower inner joint is smaller in size than the top outer joint. Adapters with joint combinations other than those listed are available.
Catalog No. 50-009-80718 Supplier Chemglass Life Sciences Supplier No. CG1002B10
Used to connect dissimilar standard taper joint sizes. Lower inner joint is smaller in size than the top outer joint. Adapters with joint combinations other than those listed are available.