Learn More
Chemglass Life Sciences Adapter, 45/50 Inner to 2" Sanitary, 316L Stainless Steel
SDP
Catalog No. 5000980719
Encompass_Preferred
Used to connect dissimilar standard taper joint sizes. Lower inner joint is smaller in size than the top outer joint. Adapters with joint combinations other than those listed are available.
Catalog No. 50-009-80719 Supplier Chemglass Life Sciences Supplier No. CG1002B22
Used to connect dissimilar standard taper joint sizes. Lower inner joint is smaller in size than the top outer joint. Adapters with joint combinations other than those listed are available.