Learn More
Chemglass Life Sciences Adapter, 45/50 Inner to 2 in Sanitary (Glass-filled PTFE) - for connecting 45/50 standard taper joints to 2 in sanitary fittings
SDP
Catalog No. 502733603
Encompass_Preferred
Glass-filled PTFE adapter that converts a 45/50 standard taper inner joint to a 2 in sanitary connection, enabling direct connection of sanitary fittings to standard taper laboratory glassware. Approximate overall length 4 in (102 mm). Clamp and seals must be purchased separately.
  • Connects 45/50 standard taper inner joints to 2 in sanitary fittings.
  • Glass-filled PTFE construction for chemical resistance and durability.
  • Approximately 4 in (102 mm) overall length.
  • Machined product line for precise fit.
  • Clamp and seals sold separately.
Catalog No. 50-273-3603 Supplier Chemglass Life Sciences Supplier No. CG1002B12
Glass-filled PTFE adapter that converts a 45/50 standard taper inner joint to a 2 in sanitary connection, enabling direct connection of sanitary fittings to standard taper laboratory glassware. Approximate overall length 4 in (102 mm). Clamp and seals must be purchased separately.
  • Connects 45/50 standard taper inner joints to 2 in sanitary fittings.
  • Glass-filled PTFE construction for chemical resistance and durability.
  • Approximately 4 in (102 mm) overall length.
  • Machined product line for precise fit.
  • Clamp and seals sold separately.