Learn More
Chemglass Life Sciences Glass Bearing Only, 45/50 Inner to #36 Internal Thread
SDP
Supplier:  Chemglass Life Sciences CG196880
Encompass_Preferred
Glass Bearing Only, 45/50 Inner to #36 Internal Thread. Bearing consists of a threaded glass adapter having a 45/50 inner joint. Design of the bearing permits replacement of the saddle o-ring at a fraction of the cost of the entire bearing.
Catalog No. CG196880