Learn More
Beckman Coulter Beckman Coulter Adapter for 30 mm conical 30-50 mL tubes, set of 4 - fits SX4750/SX4750A/JS-4.3 rotors
Catalog No. NC3257848
Encompass_Preferred
Modular disk adapters designed to hold 30-50 mL conical tubes. Sold as a set of four light green adapters for use in compatible Beckman Coulter rotors (examples: SX4750, SX4750A, JS-4.3). Autoclavable and stackable, they increase rotor flexibility and can double as storage racks.
  • Tube diameter: 30 mm.
  • Tube volume range: 30-50 mL.
  • Tube type: Conical-bottom tubes.
  • Pack quantity: set of 4 adapters.
  • Color: Light green.
  • Capacity per adapter: 4 tubes.
  • Autoclavable for routine lab use.
  • Stackable design allows use as storage racks.
Catalog No. NC3257848 Supplier Beckman Coulter Supplier No. 359154
Modular disk adapters designed to hold 30-50 mL conical tubes. Sold as a set of four light green adapters for use in compatible Beckman Coulter rotors (examples: SX4750, SX4750A, JS-4.3). Autoclavable and stackable, they increase rotor flexibility and can double as storage racks.
  • Tube diameter: 30 mm.
  • Tube volume range: 30-50 mL.
  • Tube type: Conical-bottom tubes.
  • Pack quantity: set of 4 adapters.
  • Color: Light green.
  • Capacity per adapter: 4 tubes.
  • Autoclavable for routine lab use.
  • Stackable design allows use as storage racks.