Learn More
Beckman Coulter Beckman Coulter Modular Disk Adapter for 4 x 50 mL Tubes, for GH-3.8 Rotor (359153)
Catalog No. NC2827880
Encompass_Preferred
Beckman Coulter modular disk adapter designed to hold four 50 mL round-bottom tubes and install into compatible rotor disk assemblies to enable centrifugation of larger-volume tubes.
  • Holds four 50 mL round-bottom tubes.
  • Accepts tubes up to 29 mm in diameter.
  • Seven places per adapter.
  • Designed so a full rotor holds 28 tubes.
  • rmax at adapter bottom: 184.7 mm.
  • Includes six disks per adapter assembly.
Catalog No. NC2827880 Supplier Beckman Coulter Supplier No. 359153
Beckman Coulter modular disk adapter designed to hold four 50 mL round-bottom tubes and install into compatible rotor disk assemblies to enable centrifugation of larger-volume tubes.
  • Holds four 50 mL round-bottom tubes.
  • Accepts tubes up to 29 mm in diameter.
  • Seven places per adapter.
  • Designed so a full rotor holds 28 tubes.
  • rmax at adapter bottom: 184.7 mm.
  • Includes six disks per adapter assembly.