Learn More
Beckman Coulter 500 mL conical Adapter | 4/pack
Catalog No. NC1836630
Encompass
Adapter for Tubes in Beckman Coulter Rotor used in High Performance and High Capacity Rotors: JS-5.3 AllSpin
This item is not returnable. View return policy
Catalog No. NC1836630 Supplier Beckman Coulter Supplier No. 392078
Adapter for Tubes in Beckman Coulter Rotor used in High Performance and High Capacity Rotors: JS-5.3 AllSpin