Learn More
Synthware™ Adapter, Joint to Inner Joint
GREENER_CHOICESDP
Catalog No. 31501813 Shop All Kemtech Products
Change view
Click to view available options
Product Type:
#15 O-Ring 14/20 Joint to Inner Ground Glass
#15 O-Ring 19/22 Joint to Inner Ground Glass
#15 O-Ring 24/40 Joint to Inner Ground Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
31-501-813 #15 O-Ring 24/40 Joint to Inner Ground Glass
31-501-811 #15 O-Ring 14/20 Joint to Inner Ground Glass
31-501-812 #15 O-Ring 19/22 Joint to Inner Ground Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-501-813 Supplier Synthware™ Supplier No. M131524
Add to Cart
Add to Cart
Adapter, No. 15 O-ring joint to 14/20, 19/22 or 24/40 inner joint
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass
Product Type #15 O-Ring 24/40 Joint to Inner Ground Glass
Quantity 1 Ea.
Standard Taper Joint No. 14/40