Learn More
Perkin Elmer US LLC 1⁄4 in. (6.4 mm) Internal Screw Thread Adapter with 1.8 mm O.D. Nipple, Type A
Catalog No. 509042321
Encompass_Preferred
Tubing Adapters and Connectors provide a means to connect the component shown to the screw fittings of the PTFE tubing assemblies or to connect the tubing components described. This adapter is included in the Alternate Reductant Kit (B0508594) and the Hydride Accessory Kit for Avio 200/500 (N0810433). Type A in instrument manual diagrams.
Catalog No. 50-904-2321 Supplier Perkin Elmer US LLC Supplier No. B0193342
Tubing Adapters and Connectors provide a means to connect the component shown to the screw fittings of the PTFE tubing assemblies or to connect the tubing components described. This adapter is included in the Alternate Reductant Kit (B0508594) and the Hydride Accessory Kit for Avio 200/500 (N0810433). Type A in instrument manual diagrams.