Learn More
Chemglass Life Sciences Chemglass Adapter, Air Inlet/Outlet, M27 - Stainless steel bioreactor air adapter (fits M27 headplate ports)
SDP
Catalog No. 502733304
Encompass
Stainless steel M27 air inlet/outlet adapter for bioreactors. Designed to provide overlay aeration or to serve as a gas outlet in lieu of a condenser for low aeration-rate or short-term batch applications. Constructed for corrosion resistance and a sanitary finish.
  • 316L stainless steel construction with electropolished surface (20 Ra or better).
  • M27 threaded connection for headplate ports.
  • Hose connection outer diameter: 12.00 mm.
  • Suitable for overlay aeration and low-rate gas outlet applications.
  • Compatible with common bioreactor headplate ports; replacement silicone O-ring available.
  • Designed for use in short-term batch and low-temperature processes.
Catalog No. 50-273-3304 Supplier Chemglass Life Sciences Supplier No. CLS138527
Stainless steel M27 air inlet/outlet adapter for bioreactors. Designed to provide overlay aeration or to serve as a gas outlet in lieu of a condenser for low aeration-rate or short-term batch applications. Constructed for corrosion resistance and a sanitary finish.
  • 316L stainless steel construction with electropolished surface (20 Ra or better).
  • M27 threaded connection for headplate ports.
  • Hose connection outer diameter: 12.00 mm.
  • Suitable for overlay aeration and low-rate gas outlet applications.
  • Compatible with common bioreactor headplate ports; replacement silicone O-ring available.
  • Designed for use in short-term batch and low-temperature processes.