Learn More
Chemglass Life Sciences Adapter, 14/20 Inner Joint, 0-4 valve, Airfree, Schlenk
SDP
Catalog No. AF050101
Encompass_Preferred
Adapter, 14/20 Inner Joint, 0-4 valve, Airfree, Schlenk. Adapter allows standard single neck round bottom flasks to be used as storage vessels. Sampling can be accomplished by removing 0-4mm Chem-Vac™ Chem-Cap plug while maintaining inert gas purge thru sidearm. Available with a choice of 14/20 or 24/40 standard taper inner joint.
Catalog No. AF050101 Supplier Chemglass Life Sciences Supplier No. AF050101
Adapter, 14/20 Inner Joint, 0-4 valve, Airfree, Schlenk. Adapter allows standard single neck round bottom flasks to be used as storage vessels. Sampling can be accomplished by removing 0-4mm Chem-Vac™ Chem-Cap plug while maintaining inert gas purge thru sidearm. Available with a choice of 14/20 or 24/40 standard taper inner joint.