Learn More
IDEX Health & Science Upchurch ADAPTER ASSEMBLY PEEK 10-32 FE
Catalog No. NC1873555
Encompass_Preferred
Adapter Assembly PEEK 10-32 Female x 1/4-28 Male
Catalog No. NC1873555 Supplier IDEX Health & Science Upchurch Supplier No. P669
Adapter Assembly PEEK 10-32 Female x 1/4-28 Male