Learn More
Hach Company COD Vial Adapter
Supplier:  Hach Company 4846400
Encompass
Used with Hach spectrophotometers and portable laboratories. Makes it possible to use other cells in instruments which normally take a 25-mL cell. For use with DR/800 Colorimeters.
Catalog No. 50-199-7102