Learn More
Wilmad Labglass Balloon Adapter w/PTFE Stopcock 24/40
Supplier:  Wilmad Labglass IG23T104
Encompass_Preferred
Designed with T-bore stopcock for addition or venting inert atmospheres by attaching a balloon to one of the stems. IG-22 has a 4 mm glass stopcock while IG-23T has a 4 PTFE plug.
Catalog No. 50-807-310