Learn More
Julabo USA Inc Pair, 12mm barbed fittings
SDP
Catalog No. 507534218
Encompass_Preferred
Bath Accessories, Julabo, Adapter barbed, 13.5mm OD, pair (for CD/CP/DD/MS/MX, HE/HL, SE/SL). For use with 12mm (1/2") ID rubber tubing. Requires female M16x1 nut.
Catalog No. 50-753-4218 Supplier Julabo USA Inc Supplier No. 8970445
Bath Accessories, Julabo, Adapter barbed, 13.5mm OD, pair (for CD/CP/DD/MS/MX, HE/HL, SE/SL). For use with 12mm (1/2") ID rubber tubing. Requires female M16x1 nut.