Learn More
Julabo USA Inc Pair, 8mm hose barb adapters
SDP
Catalog No. 507534219
Encompass_Preferred
Bath Accessories, Julabo, Adapter barbed, 9.5mm OD, pair (for CD/CP/DD/MS/MX, HE/HL, SE/SL). For use with 8mm ID rubber tubing. Requires M16x1 female nut to install.
Catalog No. 50-753-4219 Supplier Julabo USA Inc Supplier No. 8970446
Bath Accessories, Julabo, Adapter barbed, 9.5mm OD, pair (for CD/CP/DD/MS/MX, HE/HL, SE/SL). For use with 8mm ID rubber tubing. Requires M16x1 female nut to install.