Learn More
TA Instruments Adapter Base Mounting PCA, Mechanical Mounting Accessory, Provides Secure Installation and Alignment of PCA on Compatible DSC Instruments
Catalog No. 5000580610 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Adapter Base Mounting PCA provides secure mounting and alignment of the PCA on compatible DSC instruments. This mechanical accessory ensures proper positioning for optimal light delivery and data accuracy during photocalorimetry experiments. The base is designed for stability and ease of integration, supporting reliable performance of PCA-based DSC setups. Built for repeatable use, it enables operators to efficiently install and maintain their PCA accessory with confidence and precision.
Catalog No. 50-005-80610 Supplier TA Instruments Supplier No. 935010.901
Waters TA Instruments Adapter Base Mounting PCA provides secure mounting and alignment of the PCA on compatible DSC instruments. This mechanical accessory ensures proper positioning for optimal light delivery and data accuracy during photocalorimetry experiments. The base is designed for stability and ease of integration, supporting reliable performance of PCA-based DSC setups. Built for repeatable use, it enables operators to efficiently install and maintain their PCA accessory with confidence and precision.