Learn More
Ace Glass, Inc. Adapter, bleed, 25mm, (2)#25 Ace-Thred, glass only
SDP
Supplier:  Ace Glass, Inc. 749912
Encompass_Preferred
25mm adjustable bleed adapter used for reducing air flow in the Air Sampling Manifold to prevent a pressure drop across the sampling ports. Adapter is normally installed on front of the ACE 7511 blower, in line with the manifold. Adapter has two #25 Ace-Threds.
Catalog No. 50-974-952