Learn More
Ace Glass, Inc. Adapter, bleed, 50mm, #25, #50 Ace-Threds, glass only
SDP
Catalog No. 50974953
Encompass_Preferred
50mm adjustable bleed adapter used for reducing air flow in the Air Sampling Manifold to prevent a pressure drop across the sampling ports. Adapter is normally installed on front of the ACE 7511 blower, in line with the manifold. Adapter has one #25 Ace-Thred, and one #50 Ace-Thred.
Catalog No. 50-974-953 Supplier Ace Glass, Inc. Supplier No. 749915
50mm adjustable bleed adapter used for reducing air flow in the Air Sampling Manifold to prevent a pressure drop across the sampling ports. Adapter is normally installed on front of the ACE 7511 blower, in line with the manifold. Adapter has one #25 Ace-Thred, and one #50 Ace-Thred.