Learn More
Chemglass Life Sciences Adapter, Bushing, 24/40 Lower Inner, 10/30 Top Outer
SDP
Catalog No. CG101604
Encompass_Preferred
Adapter, Bushing, 24/40 Lower Inner, 10/30 Top Outer. Bushing style adapter having a lower standard taper inner joint and outer joint ground in the bore of the adapter. Adapters having a 10/18 or 10/30 outer member in the bore are used to insert thermometers into apparatus.
Catalog No. CG101604 Supplier Chemglass Life Sciences Supplier No. CG101604
Adapter, Bushing, 24/40 Lower Inner, 10/30 Top Outer. Bushing style adapter having a lower standard taper inner joint and outer joint ground in the bore of the adapter. Adapters having a 10/18 or 10/30 outer member in the bore are used to insert thermometers into apparatus.