Learn More
Chemglass Life Sciences Adapter, Bushing, 40/50 Lower Inner, 24/40 Top Outer
SDP
Catalog No. CG101614
Encompass_Preferred
Adapter, Bushing, 40/50 Lower Inner, 24/40 Top Outer. Bushing style adapter having a lower standard taper inner joint and outer joint ground in the bore of the adapter. Adapters having a 10/18 or 10/30 outer member in the bore are used to insert thermometers into apparatus.
Catalog No. CG101614 Supplier Chemglass Life Sciences Supplier No. CG101614
Adapter, Bushing, 40/50 Lower Inner, 24/40 Top Outer. Bushing style adapter having a lower standard taper inner joint and outer joint ground in the bore of the adapter. Adapters having a 10/18 or 10/30 outer member in the bore are used to insert thermometers into apparatus.