Learn More
Chemglass Life Sciences Adapter, Bushing, PTFE, 45/50 Lower Inner, 29/42 Top Outer
SDP
Catalog No. CG1016T06
Encompass_Preferred
Adapter, Bushing, PTFE, 45/50 Lower Inner, 29/42 Top Outer. Similar to CG1016 series of adapters, but made entirely of PTFE.
Catalog No. CG1016T06 Supplier Chemglass Life Sciences Supplier No. CG1016T06
Adapter, Bushing, PTFE, 45/50 Lower Inner, 29/42 Top Outer. Similar to CG1016 series of adapters, but made entirely of PTFE.