Learn More
Ace Glass, Inc. Adapter, filter, PTFE, #25 Ace-Thred to 1/4in female NPT, 25-50micron glass filter, CAPFE O-ring
SDP
Catalog No. 50972265
Encompass_Preferred
Catalog No. 50-972-265 Supplier Ace Glass, Inc. Supplier No. 583894