Learn More
Chemglass Life Sciences Adapter, Claisen, 14/20 Joint Size, 117mm H x 70mm W
SDP
Supplier:  Chemglass Life Sciences CG102014
Encompass_Preferred
Adapter, Claisen, 14/20 Joint Size, 117mm H x 70mm W. Outer standard taper joints on both top arms and lower inner joint. All three joints are the same size.
Catalog No. CG102014