Learn More
Chemglass Life Sciences Adapter, Claisen, 29/42 Joint, Modified
SDP
Catalog No. CG107402
Encompass_Preferred
Adapter, Claisen, 29/42 Joint, Modified. Claisen style adapter with an additional standard taper outer joint at a 45º angle to the vertical outer joint.
Catalog No. CG107402 Supplier Chemglass Life Sciences Supplier No. CG107402
Adapter, Claisen, 29/42 Joint, Modified. Claisen style adapter with an additional standard taper outer joint at a 45º angle to the vertical outer joint.