Learn More
Ace Glass, Inc. Adapter, claisen with glass stopcock
SDP
Catalog No. 50971731
Encompass_Preferred
Claisen adapter with glass stopcock. Center tube has a 10/30 outer top joint for a 76 mm immersion thermometer, and a 24/40 inner bottom joint. Adapter has a 75 degree take-off arm with a 24/40 outer top joint, and a 24/40 inner joint stopcock side arm.
Catalog No. 50-971-731 Supplier Ace Glass, Inc. Supplier No. 515010
Claisen adapter with glass stopcock. Center tube has a 10/30 outer top joint for a 76 mm immersion thermometer, and a 24/40 inner bottom joint. Adapter has a 75 degree take-off arm with a 24/40 outer top joint, and a 24/40 inner joint stopcock side arm.