Learn More
Chemglass Life Sciences Adapter, Claisen, Minum-Ware
SDP
Supplier:  Chemglass Life Sciences MW2001
Encompass_Preferred
Adapter, Claisen, Minum-Ware. Useful for addition of reagents to moisture sensitive reactions. Lower inner and top outer threaded joints all are 14/10 size. Supplied complete with two open-top screw caps (MW-102-33), PTFE faced septa (MW-104-33) and Viton o-rings (MW-100-08).
Catalog No. 50-872-922