Learn More
Chemglass Life Sciences Adapter, Connecting, 24/40 Top Outer Joint, 28/15 Lower Ball Joint
SDP
Supplier:  Chemglass Life Sciences CG100601
Encompass_Preferred
Adapter, Connecting, 24/40 Top Outer Joint, 28/15 Lower Ball Joint with top standard taper outer joint and lower spherical ball joint.
Catalog No. CG100601