Learn More
Chemglass Life Sciences Adapter, Connecting, 28/15 Top Socket, 24/40 Lower Inner
SDP
Catalog No. CG100802
Encompass_Preferred
Adapter, Connecting, 28/15 Top Socket, 24/40 Lower Inner. With top spherical socket joint and lower standard taper inner joint.
Catalog No. CG100802 Supplier Chemglass Life Sciences Supplier No. CG100802
Adapter, Connecting, 28/15 Top Socket, 24/40 Lower Inner. With top spherical socket joint and lower standard taper inner joint.