Learn More
Beckman Coulter Beckman Coulter 15 mL Conical Tube Adapter for C0650 Rotor (393088)
Catalog No. NC2596898
Encompass
The adapter allows 15 mL conical tubes to be used in rotor positions designed for 50 mL conical tubes, increasing rotor flexibility and enabling smaller-volume processing.
  • Converts rotor positions for 50 mL conical tubes to accept 15 mL conical tubes.
  • Compatible with 15 mL conical tubes up to 17 x 120 mm.
  • One adapter per unit; six adapters are required to populate a single rotor position.
  • Sold as a single adapter for installation into compatible fixed-angle rotors.
Catalog No. NC2596898 Supplier Beckman Coulter Supplier No. 393088
The adapter allows 15 mL conical tubes to be used in rotor positions designed for 50 mL conical tubes, increasing rotor flexibility and enabling smaller-volume processing.
  • Converts rotor positions for 50 mL conical tubes to accept 15 mL conical tubes.
  • Compatible with 15 mL conical tubes up to 17 x 120 mm.
  • One adapter per unit; six adapters are required to populate a single rotor position.
  • Sold as a single adapter for installation into compatible fixed-angle rotors.