Learn More
Beckman Coulter 30mm Diameter Polypropylene Tube Adapter, Quantity of One
Catalog No. NC9671774
Encompass
Adapter, 50mL Conical Tube, 30mm Maximum Tube Diameter, for TA-10-250 Fixed-Angle Rotor
This item is not returnable. View return policy
Catalog No. NC9671774 Supplier Beckman Coulter Supplier No. 356966
Adapter, 50mL Conical Tube, 30mm Maximum Tube Diameter, for TA-10-250 Fixed-Angle Rotor