Learn More
Chemglass Life Sciences 24/40 Adapter, Connecting, Height 80mm
SDP
Catalog No. 501530997
Encompass_Preferred
24/40 Adapter, Connecting, Height 80mm. Adapter has a luer outer on top and a lower standard taper inner joint.
Catalog No. 50-153-0997 Supplier Chemglass Life Sciences Supplier No. CG1055A01
24/40 Adapter, Connecting, Height 80mm. Adapter has a luer outer on top and a lower standard taper inner joint.