Learn More
Chemglass Life Sciences Adapter, Connecting, 24/40 Joint Size, 2mm Glass Stpk, Airfree, Schlenk
SDP
Catalog No. AF050002
Encompass_Preferred
Adapter, Connecting, 24/40 Joint Size, 2mm Glass Stpk, Airfree, Schlenk. Converts standard flasks and other apparatus to Airfree applications. With top outer and lower inner standard taper joints, both being the same size. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retention device or a 0-4mm Chem-Cap Airfree inlet valve. This item comes with the 2mm high vacuum glass stopcock.
Catalog No. AF050002 Supplier Chemglass Life Sciences Supplier No. AF050002
Adapter, Connecting, 24/40 Joint Size, 2mm Glass Stpk, Airfree, Schlenk. Converts standard flasks and other apparatus to Airfree applications. With top outer and lower inner standard taper joints, both being the same size. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retention device or a 0-4mm Chem-Cap Airfree inlet valve. This item comes with the 2mm high vacuum glass stopcock.