Learn More
Chemglass Life Sciences Adapter, Connecting, 24/40 Top Outer Joint, 35/20 Lower Ball Joint
SDP
Supplier:  Chemglass Life Sciences CG100602
Encompass_Preferred
Adapter, Connecting, 24/40 Top Outer Joint, 35/20 Lower Ball Joint with top standard taper outer joint and lower spherical ball joint.
Catalog No. 50-872-833