Learn More
Chemglass Life Sciences Adapter, Connecting, 10/30 Top Outer, 24/40 Lower Inner, Reducing
SDP
Supplier:  Chemglass Life Sciences CG100005
Encompass_Preferred
Adapter, Connecting, 10/30 Top Outer, 24/40 Lower Inner, Reducing. Used to connect dissimilar standard taper joint sizes. Lower inner joint is larger in size than the top outer joint.
Catalog No. 50-872-825