Learn More
Chemglass Life Sciences Adapter, Connecting, 14/20 Joint Size, 2mm Glass Stpk, Airfree, Schlenk
SDP
Catalog No. 501530962
Encompass_Preferred
Adapter, Connecting, 14/20 Joint Size, 2mm Glass Stpk, Airfree, Schlenk. Converts standard flasks and other apparatus to Airfree applications. With top outer and lower inner standard taper joints, both being the same size. Available with a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retention device.
Catalog No. 50-153-0962 Supplier Chemglass Life Sciences Supplier No. AF050001
Adapter, Connecting, 14/20 Joint Size, 2mm Glass Stpk, Airfree, Schlenk. Converts standard flasks and other apparatus to Airfree applications. With top outer and lower inner standard taper joints, both being the same size. Available with a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retention device.