Learn More
Chemglass Life Sciences Adapter, Connecting, Airfree, Schlenk, (2) 24/40 Outer Joints, (1) 24/40 Inner Joint
SDP
Catalog No. AF050402
Encompass_Preferred
Adapter, Connecting, Airfree, Schlenk, (2) 24/40 Outer Joints, (1) 24/40 Inner Joint. A 3-way adapter for the connection of 3 vessels with two top outer joints and a lower inner joint, all being the same size.
Catalog No. AF050402 Supplier Chemglass Life Sciences Supplier No. AF050402
Adapter, Connecting, Airfree, Schlenk, (2) 24/40 Outer Joints, (1) 24/40 Inner Joint. A 3-way adapter for the connection of 3 vessels with two top outer joints and a lower inner joint, all being the same size.