Learn More
Analytical Sales & Services Adapter - Converts 1/8-27 NPT Port to Accommodate 3/16" OD Tubing
Catalog No. 5000805332
Encompass
Adapter - Converts 1/8-27 NPT Port to Accommodate 3/16" OD Tubing
Catalog No. 50-008-05332 Supplier Analytical Sales & Services Supplier No. CF1827
Adapter - Converts 1/8-27 NPT Port to Accommodate 3/16" OD Tubing